Merck
  • Removal of Antibiotics from Water by Polymer of Intrinsic Microporosity: Isotherms, Kinetics, Thermodynamics, and Adsorption Mechanism.
Removal of Antibiotics from Water by Polymer of Intrinsic Microporosity: Isotherms, Kinetics, Thermodynamics, and Adsorption Mechanism.
Scientific reports (2020-01-23)
Mohammed N Alnajrani, Omar A Alsager
ABSTRACT
Traces of antibiotics within domestic and industrial effluents have toxic impact on human health as well as surrounding flora and fauna. Potential increase in antibiotic resistance of microorganisms is likely to rise due to the incomplete removal of antibiotics by traditional wastewater processing, methods such as membrane filtration and biological treatment. In this study, we investigated a novel class of material termed Polymer of Intrinsic Microporosity (PIM) that is based on amorphous microporous organic materials for the application of antibiotic removal form aqueous environments. The adsorption of four commonly used antibiotics (doxycycline, ciprofloxacin, penicillin G, and amoxicillin) was evaluated and found that at least 80% of the initial concentrations was eliminated under the optimized conditions. Langmuir and Freundlich models were then employed to correlate the equilibria data; the Freundlich model fit well the data in all cases. For kinetic data, pseudo-first and second order models were examined. Pseudo-second order model fit well the kinetic data and allowed the calculation of the adsorption rate constants. Thermodynamic parameters were obtained by conducting the adsorption studies at varied reaction temperatures. Surface potential, adsorption at various solution pHs, thermogravimetric analysis (TGA), Infrared spectroscopy (IR), and surface area experiments were conducted to draw possible adsorption mechanisms. The removal of antibiotics from water by PIM-1 is likely to be governed by both surface and pore-filling adsorption and could be facilitated by electrostatic interactions between the aromatic rings and charged functional groups as well as hydrogen bond formation between the adsorbent and adsorbate. Our work shows that the application of such novel microporous material could contribute to the removal of such challenging and persistent contaminants from wastewater with further optimizations of large-scale adsorption processes.
Sigma-Aldrich
Tetrafluoroterephthalonitrile, 99%
Pricing and availability is not currently available.