Merck
  • Pretreatment of Wheat Straw with Phosphoric Acid and Hydrogen Peroxide to Simultaneously Facilitate Cellulose Digestibility and Modify Lignin as Adsorbents.
Pretreatment of Wheat Straw with Phosphoric Acid and Hydrogen Peroxide to Simultaneously Facilitate Cellulose Digestibility and Modify Lignin as Adsorbents.
Biomolecules (2019-12-11)
Xue Wan, Fengpei Yao, Dong Tian, Fei Shen, Jinguang Hu, Yongmei Zeng, Gang Yang, Yanzong Zhang, Shihuai Deng
ABSTRACT
Effective valorization of lignin is crucial to achieve a sustainable, economic and competitive biorefinery of lignocellulosic biomass. In this work, an integrated process was proposed based on a concentrated phosphoric acid plus hydrogen peroxide (PHP) pretreatment to simultaneously facilitate cellulose digestibility and modify lignin as adsorbent. As a dominant constitutor of PHP pretreatment, H2O2 input and its influence on the overall fractionation/lignin modification performance was thoroughly investigated. Results indicated that wheat straw was fractionated more efficiently by increasing the H2O2 input. H2O2 input had a significant influence on the digestibility of the obtained cellulose-rich fraction whereby almost 100.0% cellulose-glucose conversion can be achieved even with only 0.88% H2O2 input. Besides, the adsorption capacity of lignin on MB was improved (74.3 to 210.1 mg g-1) due to the oxidative-modification in PHP pretreatment with H2O2 inputs. Regression analysis indicated that -COOH groups mainly governed the lignin adsorption (R2 = 0.946), which displayed the considerable adsorption capacities for typical cationic substances. This work shows a promising way to integrate the lignin modification concept into the emerging PHP pretreatment process with the dual goal of both cellulose utilization and lignin valorization.
Sigma-Aldrich
2-Chloro-1,3,2-dioxaphospholane, 97%