Merck
  • Suppressive Interaction Approach for Masking Stale Note of Instant Ripened Pu-Erh Tea Products.
Suppressive Interaction Approach for Masking Stale Note of Instant Ripened Pu-Erh Tea Products.
Molecules (Basel, Switzerland) (2019-12-11)
Ting Zhang, Hui Ni, Xu-Jian Qiu, Ting Li, Liang-Zhen Zhang, Li-Jun Li, Ze-Dong Jiang, Qing-Biao Li, Feng Chen, Fu-Ping Zheng
ABSTRACT
The unpleasant stale note is a negative factor hindering the consumption of instant ripened Pu-erh tea products. This study focused on investigating volatile chemicals in instant ripened Pu-erh tea that could mask the stale note via sensory evaluation, gas chromatography-mass spectrometry (GC-MS), and gas chromatography-olfactometry (GC-O) analyses. GC-MS and GC-O analyses showed that linalool, linalool oxides, trans-β-ionone, benzeneacetaldehyde, and methoxybenzenes were the major aroma contributors to the simultaneous distillation and extraction (SDE) extract of instant ripened Pu-erh tea. Sensory evaluation showed that the SDE extract had a strong stale note, which was due to methoxybenzenes. By investigating suppressive interaction among flavour components, the stale note from methoxybenzenes was shown to have reciprocal masking interactions with sweet, floral, and green notes. Moreover, the validation experiment showed that the addition of 40 μg/mL of trans-β-ionone in the instant ripened Pu-erh tea completely masked the stale note and improved the overall aromatic acceptance. These results elucidate the volatile chemicals that could mask the stale note of instant ripened Pu-erh tea products, which might help to develop high quality products made from instant ripened Pu-erh tea.
Sigma-Aldrich
Safranal, ≥90%, stabilized
Sigma-Aldrich
1,2,3-Trimethoxybenzene, 98%