Merck
  • Flow immunocytochemistry of marker expression in cells from body cavity fluids.
Flow immunocytochemistry of marker expression in cells from body cavity fluids.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2009-11-10)
Awtar Krishan, Parvin Ganjei-Azar, Ronald Hamelik, Deepti Sharma, Isildinha Reis, Mehrdad Nadji
ABSTRACT
Diagnostic cytology based on the examination of cells from body cavity fluids misses approximately 50% of patients with a proven malignancy. In an earlier study, we used immunohistochemical detection of epithelial membrane antigen expression with flow cytometric detection of DNA aneuploidy to reduce the number of false negatives. In the present study, we have combined DNA flow cytometry with flow cytometric detection of marker expression to analyze cells from body cavity fluids. Seventy-nine specimens of ascites and pleural fluids were analyzed by diagnostic cytology, DNA flow cytometry, and for the expression of the following markers: Ber-EP4, progesterone (PR), MUC4, and thyroid transcription factor-1 (TTF-1). DNA index of equal to or greater than 1.2 was seen in 33/79 (41.7%) of the samples. Statistical analysis of 79 samples in which data from cytology, DNA aneuploidy, and expression of at least one of the markers was available showed that by combining data from positive marker expression with that of aneuploidy, the sensitivity was increased from 58.5 to 100%. In contrast, out of the 38 samples designated as non-malignant by diagnostic cytology, nine had aneuploid DNA content and 16 of the diploid samples had a positive marker expression. Specificity was reduced from 74.7 to 31.6% due to the presence of aneuploidy and marker expression in these samples. ALDH1(pos)/CD44(pos)/CD24(neg) expression has been reported to be associated with human breast tumor stem cells. Some of our samples had cells with this phenotype. Flow cytometry offers the advantage of rapid multiparametric analysis of DNA aneuploidy and marker expression in cells from body cavity fluids based on the analysis of a large number of cells without observer bias. By further developing the use of specific markers and aneuploidy, it may be possible to refine flow cytometric analysis for rapid detection of malignant cells in body cavity fluids.
Sigma-Aldrich
IgG1 Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–R-Phycoerythrin antibody produced in goat, affinity isolated antibody, buffered aqueous solution