Merck
  • AhR-ROR-γt complex is a therapeutic target for MAP4K3/GLKhighIL-17Ahigh subpopulation of systemic lupus erythematosus.
AhR-ROR-γt complex is a therapeutic target for MAP4K3/GLKhighIL-17Ahigh subpopulation of systemic lupus erythematosus.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2019-07-19)
Huai-Chia Chuang, Yi-Ming Chen, Ming-Han Chen, Wei-Ting Hung, Huang-Yu Yang, Yang-Hao Tseng, Tse-Hua Tan
ABSTRACT
The cytokine IL-17A plays critical roles in the pathogenesis of autoimmune diseases. The frequencies of MAP kinase kinase kinase kinase 3 [also named germinal center kinase-like kinase (GLK)]-overexpressing T cells are correlated with disease severity of systemic lupus erythematosus (SLE). T-cell-specific GLK-transgenic mice develop spontaneous autoimmune responses through IL-17A. GLK signaling selectively stimulates IL-17A production in murine T cells through inducing aryl hydrocarbon receptor (AhR)-retinoic acid receptor-related orphan nuclear receptor-γt (ROR-γt) complex formation. Here, we investigated whether GLK-induced AhR-ROR-γt complex in T cells is a therapeutic target for human SLE. The population of GLK+IL-17A+ T cells was enhanced in the peripheral blood from patients with SLE compared with that of healthy controls using flow cytometry. The receiver operating characteristic curve analysis showed that increased GLK+IL-17A+ T-cell population in peripheral blood reflected an active stage of SLE. In addition, peripheral blood T cells from patients with SLE displayed induction of ROR-γt phosphorylation and the AhR-ROR-γt (and AhR-phosphorylated ROR-γt) complex. Moreover, we identified a small-molecule inhibitor, verteporfin, that inhibited GLK kinase activity and AhR-ROR-γt interaction. The small-molecule inhibitor verteporfin suppressed the disease severity in autoimmune mouse models and IL-17A production in T cells from patients with SLE. Collectively, the GLK-induced AhR-ROR-γt (and AhR-phosphorylated ROR-γt) complex is a therapeutic target for the GLKhighIL-17Ahigh subpopulation of human patients with SLE.-Chuang, H.-C., Chen, Y.-M., Chen, M.-H., Hung, W.-T., Yang, H.-Y., Tseng, Y.-H., Tan, T.-H. AhR-ROR-γt complex is a therapeutic target for MAP4K3/GLKhighIL-17Ahigh subpopulation of systemic lupus erythematosus.
Sigma-Aldrich
Anti-ROR gamma T Antibody, clone 6F3.1, clone 6F3.1, from mouse