Merck
  • 293 cells express both epithelial as well as mesenchymal cell adhesion molecules.
293 cells express both epithelial as well as mesenchymal cell adhesion molecules.
International journal of molecular medicine (2016-04-29)
Masakazu Inada, Genya Izawa, Wakako Kobayashi, Masayuki Ozawa
ABSTRACT
The 293 cell line, used extensively in various types of studies due to the ease with which these cells can be transfected, was thought to be derived by the transformation of primary cultures of human embryonic kidney cells with sheared adenovirus type 5 DNA. Although the 293 cells were assumed to originate from epithelial cells, the exact origin of these cells remains unknown. Previous attempts to characterize these cells combined immunostaining, immunoblot analysis and microarray analysis to demonstrate that 293 cells express neurofilament subunits, α-internexin, and several other proteins typically found in neurons. These findings raised the possibility that the 293 cell line may have originated from human neuronal lineage cells. Contrary to this suggestion, in this study, we found that the 293 cells expressed N-cadherin and vimentin, which are marker proteins expressed in mesenchymal cells. Furthermore, the 293 cells also expressed E-cadherin, cytokeratins 5/8 and desmoglein 2, which are epithelial cell markers. When the cells, primarily cultured from the kidneys of Clawn miniature swine and passaged 10-15 generations [termed porcine kidney epithelial (PKE) cells] were examined, they were found to be positive for the expression of both mesenchymal and epithelial markers. Thus, transformation by adenovirus was not necessary for the cells to express N-cadherin. Occludin and zonula occludens (ZO)-1, two components of tight junctions in epithelial and endothelial cells, were detected in the 293 and the PKE cells. Thus, the findings of the present study demonstrate that 293 cells retain several characteristics of epithelial cells.
Sigma-Aldrich
Anti-Cytokeratin 5/8, Chemicon®, from mouse
Pricing and availability is not currently available.