Merck
  • Paravertebral fascial massage promotes brain development of neonatal rats via the insulin-like growth factor 1 pathway.
Paravertebral fascial massage promotes brain development of neonatal rats via the insulin-like growth factor 1 pathway.
Neural regeneration research (2012-05-25)
Zhongqiu Wen, Wenqin Zeng, Jingxing Dai, Xin Zhou, Chun Yang, Fuhua Duan, Yufeng Liu, Huiying Yang, Lin Yuan
ABSTRACT
Massage in traditional Chinese medicine can promote body and brain development of premature and normal newborn infants. In the present study, neonatal rats (1 day old) underwent paravertebral fascial massage (15 consecutive days), followed by subcutaneous injection of insulin-like growth factor 1 receptor antagonist, JB1 (9 consecutive days). Paravertebral fascial massage significantly increased insulin-like growth factor 1 expression and cell proliferation in the subventricular zone of the lateral ventricle and dentate gyrus of the hippocampus. However, JB1 inhibited this increase. Results suggest that paravertebral fascial massage can promote brain development of neonatal rats via the insulin-like growth factor 1 pathway.
Sigma-Aldrich
Anti-BrdU Antibody, clone AH4H7-1 / 131-14871, Chemicon®, from mouse