Merck
  • Rescue of the temperature-sensitive, autosomal-recessive mutation R298S in the sodium-bicarbonate cotransporter NBCe1-A characterized by a weakened dimer and abnormal aggregation.
Rescue of the temperature-sensitive, autosomal-recessive mutation R298S in the sodium-bicarbonate cotransporter NBCe1-A characterized by a weakened dimer and abnormal aggregation.
Biochimica et biophysica acta (2015-03-07)
Harindarpal S Gill, Kun-Young Choi, Lakshmi Kammili, Anastas Popratiloff
ABSTRACT
Band keratopathy, an ocular disease that is characterized by hypercalcemia and opaque bands across the cornea, has been associated with kidney disease. Type-II renal tubular acidosis (RTA), a condition in which the kidneys fail to recover bicarbonate (HCO3-) in the proximal tubule of the nephron, results in HCO3- wastage in the urine and low blood pH. The development of these diseases is associated with autosomal-recessive mutations in the Na+-coupled HCO3- cotransporter NBCe1-A located at the basolateral membranes of either cell type. We provide insight into the devastating R298S mutation found in type-II RTA-afflicted individuals using confocal-microscopy imaging of fluorescently-tagged NBCe1-A and NBCe1-A-R298S molecules expressed in human corneal endothelial and proximal tubule cells and from in-depth biophysical studies of their cytoplasmic N-terminal domains (Nt and Nt-R298S), including Nt crystal structure, melting-temperature, and homodimer dissociation constant (KD) analyses. We illuminate and rescue trafficking defects of the R298S mutation of NBCe1-A. The KD for Nt monomer-dimer equilibrium is established. The KD for Nt-R298S is significantly higher, but immeasurable due to environmental factors (pH, temperature, concentration) that result in dimer instability leading to precipitation. The crystal structure of Nt-dimer shows that R298 is part of a putative substrate conduit and resides near the dimer interface held together by hydrogen-bond networks. The R298S is a temperature-sensitive mutation in Nt that results in instability of the colloidal system leading to abnormal aggregation. Our findings provide new perspectives to the aberrant mechanism of certain ocular pathologies and type-II RTA associated with the R298S mutation.
Sigma-Aldrich
Anti-Na+/K+ ATPase α-1 Antibody, Upstate®, from rabbit