Merck
  • α-Tocotrienol quinone modulates oxidative stress response and the biochemistry of aging.
α-Tocotrienol quinone modulates oxidative stress response and the biochemistry of aging.
Bioorganic & medicinal chemistry letters (2011-05-24)
William D Shrader, Akiko Amagata, Adam Barnes, Gregory M Enns, Andrew Hinman, Orion Jankowski, Viktoria Kheifets, Ryo Komatsuzaki, Edgar Lee, Paul Mollard, Katsuyuki Murase, Alfredo A Sadun, Martin Thoolen, Kieron Wesson, Guy Miller
ABSTRACT
We report that α-tocotrienol quinone (ATQ3) is a metabolite of α-tocotrienol, and that ATQ3 is a potent cellular protectant against oxidative stress and aging. ATQ3 is orally bioavailable, crosses the blood-brain barrier, and has demonstrated clinical response in inherited mitochondrial disease in open label studies. ATQ3 activity is dependent upon reversible 2e-redox-cycling. ATQ3 may represent a broader class of unappreciated dietary-derived phytomolecular redox motifs that digitally encode biochemical data using redox state as a means to sense and transfer information essential for cellular function.
Sigma-Aldrich
D-α-Tocotrienol, from rice, ≥75% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Vatiquinone, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Resveratrol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)