Merck
  • MYC regulation of glutamine-proline regulatory axis is key in luminal B breast cancer.
MYC regulation of glutamine-proline regulatory axis is key in luminal B breast cancer.
British journal of cancer (2017-11-24)
Madeleine L Craze, Hayley Cheung, Natasha Jewa, Nuno D M Coimbra, Daniele Soria, Rokaya El-Ansari, Mohammed A Aleskandarany, Kiu Wai Cheng, Maria Diez-Rodriguez, Christopher C Nolan, Ian O Ellis, Emad A Rakha, Andrew R Green
ABSTRACT
Altered cellular metabolism is a hallmark of cancer and some are reliant on glutamine for sustained proliferation and survival. We hypothesise that the glutamine-proline regulatory axis has a key role in breast cancer (BC) in the highly proliferative classes. Glutaminase (GLS), pyrroline-5-carboxylate synthetase (ALDH18A1), and pyrroline-5-carboxylate reductase 1 (PYCR1) were assessed at DNA/mRNA/protein levels in large, well-characterised cohorts. Gain of PYCR1 copy number and high PYCR1 mRNA was associated with Luminal B tumours. High ALDH18A1 and high GLS protein expression was observed in the oestrogen receptor (ER)+/human epidermal growth factor receptor (HER2)- high proliferation class (Luminal B) compared with ER+/HER2- low proliferation class (Luminal A) (P=0.030 and P=0.022 respectively), however this was not observed with mRNA. Cluster analysis of the glutamine-proline regulatory axis genes revealed significant associations with molecular subtypes of BC and patient outcome independent of standard clinicopathological parameters (P=0.012). High protein expression of the glutamine-proline enzymes were all associated with high MYC protein in Luminal B tumours only (P<0.001). We provide comprehensive clinical data indicating that the glutamine-proline regulatory axis plays an important role in the aggressive subclass of luminal BC and is therefore a potential therapeutic target.
Sigma-Aldrich
Anti-PYCR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ALDH18A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution