Merck
Macrophages Regulate Schwann Cell Maturation after Nerve Injury.
Cell reports (2018-09-06)
Jo Anne Stratton, Alexandra Holmes, Nicole L Rosin, Sarthak Sinha, Mohit Vohra, Nicole E Burma, Tuan Trang, Rajiv Midha, Jeff Biernaskie
ABSTRACT
Pro-regenerative macrophages are well known for their role in promoting tissue repair; however, their specific roles in promoting regeneration of the injured nerve are not well defined. Specifically, how macrophages interact with Schwann cells following injury during remyelination has been largely unexplored. We demonstrate that after injury, including in humans, macrophages function to clear debris and persist within the nerve microenvironment. Macrophage ablation immediately preceding remyelination results in an increase in immature Schwann cell density, a reduction in remyelination, and long-term deficits in conduction velocity. Targeted RNA-seq of macrophages from injured nerve identified Gas6 as one of several candidate factors involved in regulating Schwann cell dynamics. Functional studies show that the absence of Gas6 within monocyte lineage cells impairs Schwann cell remyelination within the injured nerve. These results demonstrate a role for macrophages in regulating Schwann cell function during nerve regeneration and highlight a molecular mechanism by which this occurs.
Sigma-Aldrich
Anti-O4 Antibody, clone 81, Alexa Fluor488 Conjugate, clone 81 (mAB 04), from mouse, ALEXA FLUOR 488
Sigma-Aldrich
bisBenzimide H 33258, ≥98.0% (HPLC)