Merck
  • Detection of PHLPP1α/β in human and mouse brain by different anti-PHLPP1 antibodies.
Detection of PHLPP1α/β in human and mouse brain by different anti-PHLPP1 antibodies.
Scientific reports (2015-04-02)
Travis C Jackson, Hülya Bayir, Milos D Ikonomovic, Keri Janesko-Feldman, Zaichuan Mi, Tianyan Gao, Edwin K Jackson, Patrick M Kochanek
ABSTRACT
Pleckstrin homology domain and leucine rich repeat protein phosphatase 1 (PHLPP1) is a member of the serine/threonine family of phosphatases. It has been studied in organs including brain, heart, pancreas, adipose, breast, and prostate. Human PHLPP1 encodes two splice variants - PHLPP1α (~140-150 kDa) and PHLPP1β (~180-190 kDa). Commercial antibodies are widely used to characterize PHLPP1 proteins in cells/tissues. Here we validate five different antibodies to detect PHLPP1α/β by Western blot using PHLPP1 WT/KO mice. All antibodies recognize PHLPP1β in brain. Only a single antibody (Cosmo Bio Co) detects PHLPP1α (~145-150 kDa). The other four antibodies detect a non-specific signal at ~150 kDa as evidenced by its abundance in PHLPP1 KO tissues. Results suggest Cosmo antibody is a better reagent to detect PHLPP1α by Western blot. In contrast, we found it unsuitable for immunofluorescence applications in brain. Our findings caution interpretation of the ~150 kDa band detected by some PHLPP1 antibodies in rodent and human tissues. Results also recapitulate the importance of including molecular weight standards in Western blot data to simplify retrospective analysis.
Sigma-Aldrich
Anti-PHLPP1 Antibody, from rabbit, purified by affinity chromatography