Merck
  • A novel mass assay to measure phosphatidylinositol-5-phosphate from cells and tissues.
A novel mass assay to measure phosphatidylinositol-5-phosphate from cells and tissues.
Bioscience reports (2019-10-28)
Avishek Ghosh, Sanjeev Sharma, Dhananjay Shinde, Visvanathan Ramya, Padinjat Raghu
ABSTRACT
Phosphatidylinositol-5-phosphate (PI5P) is a low abundance lipid proposed to have functions in cell migration, DNA damage responses, receptor trafficking and insulin signalling in metazoans. However, studies of PI5P function are limited by the lack of scalable techniques to quantify its level from cells and tissues in multicellular organisms. Currently, PI5P measurement requires the use of radionuclide labelling approaches that are not easily applicable in tissues or in vivo samples. In the present study, we describe a simple and reliable, non-radioactive mass assay to measure total PI5P levels from cells and tissues of Drosophila, a genetically tractable multicellular model. We use heavy oxygen-labelled ATP (18O-ATP) to label PI5P from tissue extracts while converting it into PI(4,5)P2 using an in vitro kinase reaction. The product of this reaction can be selectively detected and quantified with high sensitivity using a liquid chromatography-tandem mass spectrometry (LC-MS/MS) platform. Further, using this method, we capture and quantify the unique acyl chain composition of PI5P from Drosophila cells and tissues. Finally, we demonstrate the use of this technique to quantify elevations in PI5P levels, from Drosophila larval tissues and cultured cells depleted of phosphatidylinositol 5 phosphate 4-kinase (PIP4K), that metabolizes PI5P into PI(4,5)P2 thus regulating its levels. Thus, we demonstrate the potential of our method to quantify PI5P levels with high sensitivity from cells and tissues of multicellular organisms thus accelerating understanding of PI5P functions in vivo.
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.
Avanti
rac-16:0 PI(5)P-d5, Avanti Research - A Croda Brand 850173P, powder
Pricing and availability is not currently available.
Avanti
17:0-20:4 PI(4,5)P2, Avanti Research - A Croda Brand LM1904, powder
Pricing and availability is not currently available.
Avanti
17:0-20:4 PI(4)P, Avanti Research - A Croda Brand LM1901, powder
Pricing and availability is not currently available.