Merck
  • Diverse functions of homologous actin isoforms are defined by their nucleotide, rather than their amino acid sequence.
Diverse functions of homologous actin isoforms are defined by their nucleotide, rather than their amino acid sequence.
eLife (2017-12-16)
Pavan Vedula, Satoshi Kurosaka, Nicolae Adrian Leu, Yuri I Wolf, Svetlana A Shabalina, Junling Wang, Stephanie Sterling, Dawei W Dong, Anna Kashina
ABSTRACT
β- and γ-cytoplasmic actin are nearly indistinguishable in their amino acid sequence, but are encoded by different genes that play non-redundant biological roles. The key determinants that drive their functional distinction are unknown. Here, we tested the hypothesis that β- and γ-actin functions are defined by their nucleotide, rather than their amino acid sequence, using targeted editing of the mouse genome. Although previous studies have shown that disruption of β-actin gene critically impacts cell migration and mouse embryogenesis, we demonstrate here that generation of a mouse lacking β-actin protein by editing β-actin gene to encode γ-actin protein, and vice versa, does not affect cell migration and/or organism survival. Our data suggest that the essential in vivo function of β-actin is provided by the gene sequence independent of the encoded protein isoform. We propose that this regulation constitutes a global 'silent code' mechanism that controls the functional diversity of protein isoforms.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-beta-Actin Antibody, clone 4C2, clone 4C2, from mouse