Merck
  • A Modular Probe Strategy for Drug Localization, Target Identification and Target Occupancy Measurement on Single Cell Level.
A Modular Probe Strategy for Drug Localization, Target Identification and Target Occupancy Measurement on Single Cell Level.
ACS chemical biology (2016-07-08)
Anna Rutkowska, Douglas W Thomson, Johanna Vappiani, Thilo Werner, Katrin M Mueller, Lars Dittus, Jana Krause, Marcel Muelbaier, Giovanna Bergamini, Marcus Bantscheff
ABSTRACT
Late stage failures of candidate drug molecules are frequently caused by off-target effects or inefficient target engagement in vivo. In order to address these fundamental challenges in drug discovery, we developed a modular probe strategy based on bioorthogonal chemistry that enables the attachment of multiple reporters to the same probe in cell extracts and live cells. In a systematic evaluation, we identified the inverse electron demand Diels-Alder reaction between trans-cyclooctene labeled probe molecules and tetrazine-tagged reporters to be the most efficient bioorthogonal reaction for this strategy. Bioorthogonal biotinylation of the probe allows the identification of drug targets in a chemoproteomics competition binding assay using quantitative mass spectrometry. Attachment of a fluorescent reporter enables monitoring of spatial localization of probes as well as drug-target colocalization studies. Finally, direct target occupancy of unlabeled drugs can be determined at single cell resolution by competitive binding with fluorescently labeled probe molecules. The feasibility of the modular probe strategy is demonstrated with noncovalent PARP inhibitors.
Sigma-Aldrich
Biotin-PEG4-alkyne, for copper catalyzed click labeling