Merck
  • Cell-Cycle Asynchrony Generates DNA Damage at Mitotic Entry in Polyploid Cells.
Cell-Cycle Asynchrony Generates DNA Damage at Mitotic Entry in Polyploid Cells.
Current biology : CB (2019-11-12)
Maddalena Nano, Simon Gemble, Anthony Simon, Carole Pennetier, Vincent Fraisier, Veronique Marthiens, Renata Basto
ABSTRACT
Polyploidy arises from the gain of complete chromosome sets [1], and it is known to promote cancer genome evolution. Recent evidence suggests that a large proportion of human tumors experience whole-genome duplications (WGDs), which might favor the generation of highly abnormal karyotypes within a short time frame, rather than in a stepwise manner [2-6]. However, the molecular mechanisms linking whole-genome duplication to genetic instability remain poorly understood. Using repeated cytokinesis failure to induce polyploidization of Drosophila neural stem cells (NSCs) (also called neuroblasts [NBs]), we investigated the consequences of polyploidy in vivo. Surprisingly, we found that DNA damage is generated in a subset of nuclei of polyploid NBs during mitosis. Importantly, our observations in flies were confirmed in mouse NSCs (mNSCs) and human cancer cells after acute cytokinesis inhibition. Interestingly, DNA damage occurs in nuclei that were not ready to enter mitosis but were forced to do so when exposed to the mitotic environment of neighboring nuclei within the same cell. Additionally, we found that polyploid cells are cell-cycle asynchronous and forcing cell-cycle synchronization was sufficient to lower the levels of DNA damage generated during mitosis. Overall, this work supports a model in which DNA damage at mitotic entry can generate DNA structural abnormalities that might contribute to the onset of genetic instability.
Sigma-Aldrich
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, ≥97.0% (AT)
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-phospho-Histone H3 (pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Supelco
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid, pharmaceutical impurity standard