Merck
  • Identification and functional response of interstitial Cajal-like cells from rat mesenteric artery.
Identification and functional response of interstitial Cajal-like cells from rat mesenteric artery.
Cell and tissue research (2011-01-19)
Aurélie Formey, Lara Buscemi, François-Xavier Boittin, Jean-Louis Bény, Jean-Jacques Meister
ABSTRACT
Cells with irregular shapes, numerous long thin filaments, and morphological similarities to the gastrointestinal interstitial cells of Cajal (ICCs) have been observed in the wall of some blood vessels. These ICC-like cells (ICC-LCs) do not correspond to the other cell types present in the arterial wall: smooth muscle cells (SMCs), endothelial cells, fibroblasts, inflammatory cells, or pericytes. However, no clear physiological role has as yet been determined for ICC-LCs in the vascular wall. The aim of this study has been to identify and characterize the functional response of ICC-LCs in rat mesenteric arteries. We have observed ICC-LCs and identified them morphologically and histologically in three different environments: isolated artery, freshly dispersed cells, and primary-cultured cells from the arterial wall. Like ICCs but unlike SMCs, ICC-LCs are positively stained by methylene blue. Cells morphologically resembling methylene-blue-positive cells are also positive for the ICC and ICC-LC markers α-smooth muscle actin and desmin. Furthermore, the higher expression of vimentin in ICC-LCs compared with SMCs allows a clear discrimination between these two cell types. At the functional level, the differences observed in the variations of cytosolic free calcium concentration of freshly dispersed SMCs and ICC-LCs in response to a panel of vasoactive molecules show that ICC-LCs, unlike SMCs, do not respond to exogenous ATP and [Arginine](8)-vasopressin.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Sigma-Aldrich
Angiotensin II, Human, Plays an important role in the regulation of blood pressure.
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Elastase from porcine pancreas, Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)
Sigma-Aldrich
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 IU/mL in 0.9% NaCl
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥97% (HPLC), crystalline, from microbial
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission