Merck
  • Synthesis of Chiral 1,4-Benzodioxanes and Chromans by Enantioselective Palladium-Catalyzed Alkene Aryloxyarylation Reactions.
Synthesis of Chiral 1,4-Benzodioxanes and Chromans by Enantioselective Palladium-Catalyzed Alkene Aryloxyarylation Reactions.
Angewandte Chemie (International ed. in English) (2016-03-19)
Naifu Hu, Ke Li, Zheng Wang, Wenjun Tang
ABSTRACT
A highly enantioselective alkene aryloxyarylation led to the high-yielding formation of a series of 1,4-benzodioxanes, 1,4-benzooxazines, and chromans containing quaternary stereocenters with excellent enantioselectivity. The sterically bulky and conformationally well defined chiral monophosphorus ligand L4 or L5 was responsible for the high reactivity and enantioselectivity of these transformations. The application of this method to the synthesis of the chiral chroman backbone of α-tocopherol was demonstrated.
Sigma-Aldrich
(R)-AntPhos, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
(S)-AntPhos, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
(R)-BIDIME, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
(S)-BIDIME, ≥97%
Pricing and availability is not currently available.