Merck
  • Proteomic Phosphosite Analysis Identified Crucial NPM-ALK-Mediated NIPA Serine and Threonine Residues.
Proteomic Phosphosite Analysis Identified Crucial NPM-ALK-Mediated NIPA Serine and Threonine Residues.
International journal of molecular sciences (2019-08-23)
Anina Gengenbacher, Alina Müller-Rudorf, Teresa Poggio, Linda Gräßel, Veronica I Dumit, Stefanie Kreutmair, Lena J Lippert, Justus Duyster, Anna L Illert
ABSTRACT
Anaplastic large-cell lymphoma (ALCL) is an aggressive non-Hodgkin lymphoma that shows in 60% of cases a translocation t(2;5)(p23;q35), which leads to the expression of the oncogenic kinase NPM-ALK. The nuclear interaction partner of ALK (NIPA) defines an E3-SCF ligase that contributes to the timing of mitotic entry. It has been shown that co-expression of NIPA and NPM-ALK results in constitutive NIPA phosphorylation. By mass spectrometry-based proteomics we identified nine serine/threonine residues to be significantly upregulated in NIPA upon NPM-ALK expression. Generation of phospho-deficient mutants of the respective phospho-residues specified five serine/threonine residues (Ser-338, Ser-344, Ser-370, Ser-381 and Thr-387) as key phosphorylation sites involved in NPM-ALK-directed phosphorylation of NIPA. Analysis of the biological impact of NIPA phosphorylation by NPM-ALK demonstrated that the ALK-induced phosphorylation does not change the SCFNIPA-complex formation but may influence the localization of NIPA and NPM-ALK. Biochemical analyses with phospho-deficient mutants elucidated the importance of NIPA phosphorylation by NPM-ALK for the interaction of the two proteins and proliferation potential of respective cells: Silencing of the five crucial NIPA serine/threonine residues led to a highly enhanced NIPA-NPM-ALK binding capacity as well as a slightly reduced proliferation in Ba/F3 cells.
Sigma-Aldrich
Anti-ZC3HC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid