Merck
  • Caspase-11-dependent IL-1α release boosts Th17 immunity against Paracoccidioides brasiliensis.
Caspase-11-dependent IL-1α release boosts Th17 immunity against Paracoccidioides brasiliensis.
PLoS pathogens (2019-08-20)
Natália Ketelut-Carneiro, Camila Oliveira Silva Souza, Luciana Benevides, Luiz Gustavo Gardinassi, Maria Cláudia Silva, Lucas Alves Tavares, Dario Simões Zamboni, João Santana Silva
ABSTRACT
The granulomatous lesion resulting from infection with the fungus Paracoccidioides brasiliensis is characterized by a compact aggregate of mature cells, surrounded by a fibroblast- and collagen-rich content. Granuloma formation requires signaling elicited by inflammatory molecules such as members of the interleukin-1 family. Two members of this family have been thoroughly studied, namely IL-1α and IL-1β. In this study, we addressed the mechanisms underlying IL-1α secretion and its functional role on the host resistance to fungal infection. We found that, the expression of caspase-11 triggered by P. brasiliensis infection of macrophages depends on IFN-β production, because its inhibition reduced procaspase-11 levels. Curiously, caspase-11 deficiency did not impair IL-1β production, however caspase-11 was required for a rapid pore-mediated cell lysis. The plasma membrane rupture facilitated the release of IL-1α, which was necessary to induce NO production and restrict fungal replication. Furthermore, P. brasiliensis-infected macrophages required IL-1α to produce optimal levels of IL-6, a major component of Th17 lymphocyte differentiation. Indeed, IL-1α deficiency accounted for a significant reduction of Th17 lymphocytes in lungs of infected mice, correlating with diminished neutrophil infiltration in the lungs. Strikingly, we identified that IL-1α directly reprograms the transcriptional profile of Th17-committed lymphocytes, increasing cellular proliferation, as for boosting IL-17 production by these cells. Beyond neutrophil chemotaxis in vivo, IL-17 also amplified IL-1α production by infected macrophages in vitro, endorsing a critical amplification loop of the inflammatory response. Therefore, our data suggest that the IFN-β/caspase-11/IL-1α pathway shapes a protective antifungal Th17 immunity, revealing a molecular mechanism underlying the cross-talk between innate and adaptive immunity.