Merck
  • Phenolic compounds from coffee by-products modulate adipogenesis-related inflammation, mitochondrial dysfunction, and insulin resistance in adipocytes, via insulin/PI3K/AKT signaling pathways.
Phenolic compounds from coffee by-products modulate adipogenesis-related inflammation, mitochondrial dysfunction, and insulin resistance in adipocytes, via insulin/PI3K/AKT signaling pathways.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2019-07-16)
Miguel Rebollo-Hernanz, Qiaozhi Zhang, Yolanda Aguilera, Maria A Martín-Cabrejas, Elvira Gonzalez de Mejia
ABSTRACT
The aim of this study was to evaluate the inhibitory potential of aqueous extracts from coffee silverskin (CSE) and husk (CHE) and their main phenolics on adipogenesis, obesity-related inflammation, mitochondrial dysfunction, and insulin resistance, in vitro. Coffee by-products extracts (31-500 μg mL-1) and pure phenolics (100 μmol L-1) reduced lipid accumulation and increased mitochondrial activity in 3T3-L1 adipocytes. Also reduced the expression of inducible nitric oxide synthase and cyclooxygenase-2 and diminished secretion of pro-inflammatory factors in LPS-stimulated RAW2643.7 macrophages. Cytokine release diminished (tumor necrosis factor α: 23-57%; monocyte chemoattractant protein 1: 42-60%; interleukin-6: 30-39%) and adiponectin increased (7-13- fold) in adipocytes treated with macrophage-conditioned media. ROS scavenging and activation of peroxisome proliferator-activated receptor γ coactivator 1-α pathway counteracted mitochondrial dysfunction. Increases in insulin receptor (1.4 to 4-fold), phosphoinositide 3-kinase (2 to 3-fold) and protein kinase B (1.3 to 3-fold) phosphorylation, in conjunction with a decrease in serine phosphorylation of insulin receptor substrate 1, evoked glucose transporter 4 translocation (8-15-fold) and glucose uptake (44-85%). CSE and CHE phenolics inhibited adipogenesis and elicited adipocytes browning. Suppressing macrophages-adipocytes interaction alleviated inflammation-triggered mitochondrial dysfunction and insulin resistance. CSE and CHE are beneficial in reducing adipogenesis and inflammation-related disorders.
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzoic-2,3,5,6-d4 acid, 98 atom % D
Pricing and availability is not currently available.