Merck
  • Application of Dolichos biflorus in immunoassay detection of kidney collecting duct biomarkers.
Application of Dolichos biflorus in immunoassay detection of kidney collecting duct biomarkers.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2010-05-25)
Samer Sourial, Lydia Searchfield, Ina Schuppe-Koistinen, Graham R Betton, Daniela Riccardi, Sally A Price
ABSTRACT
Currently there are no biomarkers for detecting collecting duct damage in man. Antibodies to several collecting duct-specific antigens exist but sandwich assays have been difficult to establish due to the need for two different antibodies to the same protein. We hypothesized that a collecting duct-specific lectin could be used in combination with a collecting duct-specific antibody to negate the need for two different antibodies. The collecting duct specificity of selected antibodies (NiCa II 13C2, Pap XI 3C7, HuPaP VII 2B11 and aquaporin 2), was verified by immunohistochemistry. Aquaporin 2 and Pap XI 3C7 were used successfully in setting up assays with the lectin Dolichos biflorus, using the Meso Scale Discovery (MSD) platform. Antigen expression was highest in the papillae of rat and human kidney (corresponding to the greatest density of collecting ducts) and was also present in normal urine. We propose that further qualification and validation would lead to an assay for detecting collecting duct damage in man.
Sigma-Aldrich
2-Bromoethylamine hydrobromide, purum, ≥97.0% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Bromoethylamine hydrobromide, 99%
Pricing and availability is not currently available.