Merck
  • Inactivation of nuclear histone deacetylases by EP300 disrupts the MiCEE complex in idiopathic pulmonary fibrosis.
Inactivation of nuclear histone deacetylases by EP300 disrupts the MiCEE complex in idiopathic pulmonary fibrosis.
Nature communications (2019-05-22)
Karla Rubio, Indrabahadur Singh, Stephanie Dobersch, Pouya Sarvari, Stefan Günther, Julio Cordero, Aditi Mehta, Lukasz Wujak, Hector Cabrera-Fuentes, Cho-Ming Chao, Peter Braubach, Saverio Bellusci, Werner Seeger, Andreas Günther, Klaus T Preissner, Malgorzata Wygrecka, Rajkumar Savai, Dulce Papy-Garcia, Gergana Dobreva, Mathias Heikenwalder, Soni Savai-Pullamsetti, Thomas Braun, Guillermo Barreto
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a chronic, progressive, and highly lethal lung disease with unknown etiology and poor prognosis. IPF patients die within 2 years after diagnosis mostly due to respiratory failure. Current treatments against IPF aim to ameliorate patient symptoms and to delay disease progression. Unfortunately, therapies targeting the causes of or reverting IPF have not yet been developed. Here we show that reduced levels of miRNA lethal 7d (MIRLET7D) in IPF compromise epigenetic gene silencing mediated by the ribonucleoprotein complex MiCEE. In addition, we find that hyperactive EP300 reduces nuclear HDAC activity and interferes with MiCEE function in IPF. Remarkably, EP300 inhibition reduces fibrotic hallmarks of in vitro (patient-derived primary fibroblast), in vivo (bleomycin mouse model), and ex vivo (precision-cut lung slices, PCLS) IPF models. Our work provides the molecular basis for therapies against IPF using EP300 inhibition.
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
UNC1999, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Fibronectin Antibody, Chemicon®, from rabbit