Merck
  • A liquid-like spindle domain promotes acentrosomal spindle assembly in mammalian oocytes.
A liquid-like spindle domain promotes acentrosomal spindle assembly in mammalian oocytes.
Science (New York, N.Y.) (2019-06-30)
Chun So, K Bianka Seres, Anna M Steyer, Eike Mönnich, Dean Clift, Anastasija Pejkovska, Wiebke Möbius, Melina Schuh
ABSTRACT
Mammalian oocytes segregate chromosomes with a microtubule spindle that lacks centrosomes, but the mechanisms by which acentrosomal spindles are organized and function are largely unclear. In this study, we identify a conserved subcellular structure in mammalian oocytes that forms by phase separation. This structure, which we term the liquid-like meiotic spindle domain (LISD), permeates the spindle poles and forms dynamic protrusions that extend well beyond the spindle. The LISD selectively concentrates multiple microtubule regulatory factors and allows them to diffuse rapidly within the spindle volume. Disruption of the LISD via different means disperses these factors and leads to severe spindle assembly defects. Our data suggest a model whereby the LISD promotes meiotic spindle assembly by serving as a reservoir that sequesters and mobilizes microtubule regulatory factors in proximity to spindle microtubules.
Sigma-Aldrich
Anti-PCM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-MCRS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-KANSL3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CEP164 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SSX2IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TGN46, Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Anti-Topoisomerase II Antibody, clone KiS1, clone KiS1, Chemicon®, from mouse