Merck
  • Controlled Synthesis of Block Copolymers by Mechanistic Transformation from Atom Transfer Radical Polymerization to Iniferter Process.
Controlled Synthesis of Block Copolymers by Mechanistic Transformation from Atom Transfer Radical Polymerization to Iniferter Process.
Macromolecular rapid communications (2019-05-16)
Cansu Aydogan, Mustafa Ciftci, Yusuf Yagci
ABSTRACT
A straightforward transformation protocol combining two distinct living polymerization methods for the controlled synthesis of block copolymers is described. In the first step, bromo-terminated poly(methyl methacrylate) is prepared by atom transfer radical polymerization (ATRP). Then, a bromide end group is substituted with a triphenylmethyl (trityl) functionality under visible light irradiation using dimanganese decacarbonyl (Mn2 (CO)10 ) photochemistry. The resulting polymers with trityl end groups are used as macroiniferter for the polymerization of styrene and tert-butyl acrylate (tBA) to yield desired block copolymers with narrow molecular weight distribution. Moreover, the amphiphilic copolymers with acrylic acid functionalities are obtained by the hydrolyzation of poly(tert-butyl acrylate) containing block copolymers with trifluoroacetic acid.