Merck
  • Augmented improvement of cognition and memory by aripiprazole add-on for cilostazol treatment in the chronic cerebral hypoperfusion mouse model.
Augmented improvement of cognition and memory by aripiprazole add-on for cilostazol treatment in the chronic cerebral hypoperfusion mouse model.
Behavioural brain research (2019-03-10)
So Youn Park, Hae Young Kim, Yi Sle Lee, Hye Jin Heo, Hwa Kyoung Shin, Won Suk Lee, Ki Whan Hong, Chi Dae Kim
ABSTRACT
Cerebrovascular dysfunction is associated with cognitive impairment in vascular dementia patients. This study aimed to explore augmented improvement of cognition and memory by aripiprazole add-on for cilostazol treatment in vascular dementia model. Male C57BL/6 mice were subjected to BCAS, and spatial probe and memory retention were examined using the Morris water maze (MWM) test. In the present study, the escape latency on the first day after 3rd week was 21.4 ± 4.0 s in sham-operated mice, and 76.3 ± 4.2 s in the vehicle-treated BCAS mice. In the spatial probe tests in the 3rd week, aripiprazole (1 mg/kg/day) showed time-dependently amelioration in spatial learning and memory impairments in contrast to 0.5 mg/kg/day. After treatment with 20 mg/kg/day of cilostazol for 3 weeks, the escape latency significantly decreased to 26.6 ± 5.8 s on the first day and further shortened to 21.6 ± 6.8 s on the fourth day. When the BCAS mice were concurrently treated with 0.5 mg/kg/day aripiprazole plus 20 mg/kg/day of cilostazol for 3 weeks, the escape latency was more shortened from 20.4 ± 1.2 s (1st day) to 14.9 ± 1.7 s on the 4th day of the 3-week trials. Furthermore, decreased spatial memory retention in BCAS mice was significantly alleviated by aripiprazole plus cilostazol cotreatment, indicating the benefit of aripiprazole add-on therapy. In line with these, significantly increased mBDNF and P-CREB levels and reduced apoptosis were identified in the BCAS mouse brain dentate gyrus by cotreatment as contrasted to each monotherapy. These results may provide the synergistic therapeutic avenues for augmented improvement of cognition and memory by cotreatment with aripiprazole plus cilostazol in cases of vascular dementia.