Merck
  • RABL2 positively controls localization of GPCRs in mammalian primary cilia.
RABL2 positively controls localization of GPCRs in mammalian primary cilia.
Journal of cell science (2018-12-24)
Izumi Dateyama, Yoshihiro Sugihara, Shuhei Chiba, Reo Ota, Risa Nakagawa, Tetsuo Kobayashi, Hiroshi Itoh
ABSTRACT
The primary cilium, a solitary protrusion from most mammalian cells, functions as a cell sensor by receiving extracellular signals through receptors and channels accumulated in the organelle. Certain G-protein-coupled receptors (GPCRs) specifically localize to the membrane compartment of primary cilia. To gain insight into the mechanisms that regulate ciliary GPCR sorting, we investigated the atypical small GTPase RAB-like 2 (RABL2; herein referring to the near-identical human paralogs RABL2A and RABL2B). RABL2 recruitment to the mother centriole is dependent on the distal appendage proteins CEP164 and CEP83. We found that silencing of RABL2 causes mis-targeting of ciliary GPCRs, GPR161 and HTR6, whereas overexpression of RABL2 resulted in accumulation of these receptors in the organelle. Ablation of CEP19 and the intraflagellar transport B (IFT-B) complex, which interact with RABL2, also leads to mis-localization of GPR161. RABL2 controls localization of GPR161 independently of TULP3, which promotes entry of ciliary GPCRs. We further demonstrate that RABL2 physically associates with ciliary GPCRs. Taken together, these studies suggest that RABL2 plays an important role in trafficking of ciliary GPCRs at the ciliary base in mammalian cells.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-Ninein Antibody, clone 79-160-7, clone 79-160-7, from mouse