- Operando Study Reveals the Superior Cracking Activity and Stability of Hierarchical ZSM-5 Catalyst for the Cracking of Low-Density Polyethylene.
Operando Study Reveals the Superior Cracking Activity and Stability of Hierarchical ZSM-5 Catalyst for the Cracking of Low-Density Polyethylene.
ChemSusChem (2018-12-07)
Karolina A Tarach, Kamila Pyra, Sandra Siles, Ignacio Melián-Cabrera, Kinga Góra-Marek
PMID30520260
ABSTRACT
A new theoretical and practical framework has been developed through operando study of the zeolite catalytic cracking of low-density polyethylene (as a model reaction) under reaction conditions. Results show that microporous ZSM-5 gives rise to less cracking products. Hierarchical ZSM-5 zeolites are more active cracking catalysts, rendering more C2 -C5 hydrocarbons, with a delayed deactivation due to the secondary porosity. This tool in combination with thermogravimetric analysis provides complementary and valuable information for the study, and design of advanced catalysts.