Merck
  • Leucine Promotes Milk Synthesis in Bovine Mammary Epithelial Cells via the PI3K-DDX59 Signaling.
Leucine Promotes Milk Synthesis in Bovine Mammary Epithelial Cells via the PI3K-DDX59 Signaling.
Journal of agricultural and food chemistry (2019-07-28)
Youwen Qiu, Bo Qu, Zhen Zhen, Xiaohan Yuan, Li Zhang, Minghui Zhang
ABSTRACT
Leucine is an essential amino acid in the milk production of bovine mammary glands, but the regulatory roles and molecular mechanisms of leucine are still not known well. This study investigated the roles of leucine on milk synthesis and explored the corresponding mechanism in bovine mammary epithelial cells (BMECs). Leucine (0, 0.25, 0.5, 0.75, 1.0, and 1.25 mM) was added to BMECs that were cultured in FBS-free OPTI-MEM medium. Leucine significantly promoted milk protein and milk fat synthesis and also increased phosphorylation of mTOR signaling protein and the protein expression levels of SREBP-1c, with the most significant effects at 0.75 mM concentration. Leucine increased the expression and nuclear localization of DDX59, and loss and gain of gene function experiments further reveal that DDX59 mediates the stimulation of leucine on the mRNA expression variation of mTOR and SREBP-1c genes. PI3K inhibition experiment further detected that leucine upregulated expression of DDX59 and its downstream signaling via PI3K activation. ChIP-qPCR analysis further proved the binding of DDX59 to the promoter regions of mTOR and SREBP-1c. In summary, these data prove that DDX59 positively regulates the mTOR and SREBP-1c signaling pathways leading to synthesis of milk, and leucine regulates these two signaling pathways through the PI3K-DDX59 signaling.