- Biosynthesis of O-N-acetylglucosamine-linked glycans in Trypanosoma cruzi. Characterization of the novel uridine diphospho-N-acetylglucosamine:polypeptide N-acetylglucosaminyltransferase-catalyzing formation of N-acetylglucosamine alpha1-->O-threonine.
Biosynthesis of O-N-acetylglucosamine-linked glycans in Trypanosoma cruzi. Characterization of the novel uridine diphospho-N-acetylglucosamine:polypeptide N-acetylglucosaminyltransferase-catalyzing formation of N-acetylglucosamine alpha1-->O-threonine.
In this study, we have characterized the activity of a uridine diphospho-N-acetylglucosamine:polypeptide-alpha-N-acetylglucosaminylt ransferase (O-alpha-GlcNAc-transferase) from Trypanosoma cruzi. The activity is present in microsomal membranes and is responsible for the addition of O-linked alpha-N-acetylglucosamine to cell surface proteins. This preparation adds N-acetylglucosamine to a synthetic peptide KPPTTTTTTTTKPP containing the consensus threonine-rich dodecapeptide encoded by T. cruzi MUC gene (Di Noia, J. M., Sánchez D. O., and Frasch, A. C. C. (1995) J. Biol. Chem. 270, 24146-24149). Incorporation of N-[3H]acetylglucosamine is linearly dependent on incubation time and concentration of enzyme and substrate. The transferase activity has an optimal pH of 7.5- 8.5, requires Mn2+, is unaffected by tunicamycin or amphomycin, and is strongly inhibited by UDP. The optimized synthetic peptide acceptor for the cytosolic O-GlcNAc-transferase (YSDSPSTST) (Haltiwanger, R. S., Holt, G. D., and Hart, G. W. (1990) J. Biol. Chem. 265, 2563-2568) is not a substrate for this enzyme. The glycosylated KPPTTTTTTTTKPP product is susceptible to base-catalyzed beta-elimination, and the presence of N-acetylglucosamine alpha-linked to threonine is supported by enzymatic digestion and nuclear magnetic resonance data. These results describe a unique biosynthetic pathway for T. cruzi surface mucin-like molecules, with potential chemotherapeutic implications.