Merck
  • Nystatin induces secretion of interleukin (IL)-1beta, IL-8, and tumor necrosis factor alpha by a toll-like receptor-dependent mechanism.
Nystatin induces secretion of interleukin (IL)-1beta, IL-8, and tumor necrosis factor alpha by a toll-like receptor-dependent mechanism.
Antimicrobial agents and chemotherapy (2005-07-29)
Raymund R Razonable, Martin Henault, Harold L Watson, Carlos V Paya
ABSTRACT
Nystatin is an antifungal compound with potent proinflammatory properties. Herein, we demonstrate that nystatin induces interleukin (IL)-1beta, IL-8, and tumor necrosis factor alpha secretion through its activation of toll-like receptor 1 (TLR1) and TLR2. Hence, a TLR-dependent mechanism could serve as the molecular basis for the proinflammatory properties of nystatin.
Sigma-Aldrich
Nystatin, ≥4,400 units/mg
Pricing and availability is not currently available.