Merck
  • Transforming growth factor-beta receptor inhibition enhances adenoviral infectability of carcinoma cells via up-regulation of Coxsackie and Adenovirus Receptor in conjunction with reversal of epithelial-mesenchymal transition.
Transforming growth factor-beta receptor inhibition enhances adenoviral infectability of carcinoma cells via up-regulation of Coxsackie and Adenovirus Receptor in conjunction with reversal of epithelial-mesenchymal transition.
Cancer research (2006-02-03)
Markus D Lacher, Maarit I Tiirikainen, Elise F Saunier, Christine Christian, Mario Anders, Martin Oft, Allan Balmain, Rosemary J Akhurst, Wolfgang Michael Korn
ABSTRACT
Expression of the Coxsackie and Adenovirus Receptor (CAR) is frequently reduced in carcinomas, resulting in decreased susceptibility of such tumors to infection with therapeutic adenoviruses. Because CAR participates physiologically in the formation of tight-junction protein complexes, we examined whether molecular mechanisms known to down-regulate cell-cell adhesions cause loss of CAR expression. Transforming growth factor-beta (TGF-beta)-mediated epithelial-mesenchymal transition (EMT) is a phenomenon associated with tumor progression that is characterized by loss of epithelial-type cell-cell adhesion molecules (including E-cadherin and the tight junction protein ZO-1), gain of mesenchymal biochemical markers, such as fibronectin, and acquisition of a spindle cell phenotype. CAR expression is reduced in tumor cells that have undergone EMT in response to TGF-beta. This down-regulation results from repression of CAR gene transcription, whereas altered RNA stability and increased proteasomal protein degradation play no role. Loss of CAR expression in response to TGF-beta is accompanied by reduced susceptibility to adenovirus infection. Indeed, treatment of carcinoma cells with LY2109761, a specific pharmacologic inhibitor of TGF-beta receptor types I and II kinases, resulted in increased CAR RNA and protein levels as well as improved infectability with adenovirus. This was observed in cells induced to undergo EMT by addition of exogenous TGF-beta and in those that were transformed by endogenous autocrine/paracrine TGF-beta. These findings show down-regulation of CAR in the context of EMT and suggest that combination of therapeutic adenoviruses and TGF-beta receptor inhibitors could be an efficient anticancer strategy.
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Lactacystin, Synthetic, Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC₅₀ = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.
Sigma-Aldrich
LY2109761, ≥98% (HPLC)