Merck
  • Purification and cDNA cloning of a cellulase from abalone Haliotis discus hannai.
Purification and cDNA cloning of a cellulase from abalone Haliotis discus hannai.
European journal of biochemistry (2003-02-13)
Ken-ichi Suzuki, Takao Ojima, Kiyoyoshi Nishita
ABSTRACT
A cellulase [endo-beta-1,4-D-glucanase (EC 3.2.1.4)] was isolated from the hepatopancreas of abalone Haliotis discus hannai by successive chromatographies on TOYOPEARL CM-650M, hydroxyapatite and Sephacryl S-200 HR. The molecular mass of the cellulase was estimated to be 66 000 Da by SDS/PAGE, thus the enzyme was named HdEG66. The hydrolytic activity of HdEG66 toward carboxymethylcellulose showed optimal temperature and pH at 38 degrees C and 6.3, respectively. cDNAs encoding HdEG66 were amplified by the polymerase chain reaction from an abalone hepatopancreas cDNA library with primers synthesized on the basis of partial amino-acid sequences of HdEG66. By overlapping the nucleotide sequences of the cDNAs, a sequence of 1898 bp in total was determined. The coding region of 1785 bp located at nucleotide position 56-1840 gave an amino-acid sequence of 594 residues including the initiation methionine. The N-terminal region of 14 residues in the deduced sequence was regarded as the signal peptide as it was absent in HdEG66 protein and showed high similarity to the consensus sequence for signal peptides of eukaryote secretory proteins. Thus, matured HdEG66 was thought to consist of 579 residues. The C-terminal region of 453 residues in HdEG66, i.e. approximately the C-terminal three quarters of the protein, showed 42-44% identity to the catalytic domains of glycoside hydrolase family 9 (GHF9)-cellulases from arthropods and Thermomonospora fusca. While the N-terminal first quarter of HdEG66 showed 27% identity to the carbohydrate-binding module (CBM) of a Cellulomonas fimi cellulase, CenA. Thus, the HdEG66 was regarded as the GHF9-cellulase possessing a family II CBM in the N-terminal region. By genomic PCR using specific primers to the 3'-terminal coding sequences of HdEG66-cDNA, a DNA of 2186 bp including three introns was amplified. This strongly suggests that the origin of HdEG66 is not from symbiotic bacteria but abalone itself.
Product Description
TOYOPEARL® CM-650M Bulk Media, phase carboxymethyl, bottle of 250 mL, 65 μm particle size