Merck
  • Phosphatidylinositol 3'-kinase and p70s6k are required for insulin but not bisperoxovanadium 1,10-phenanthroline (bpV(phen)) inhibition of insulin-like growth factor binding protein gene expression. Evidence for MEK-independent activation of mitogen-activated protein kinase by bpV(phen).
Phosphatidylinositol 3'-kinase and p70s6k are required for insulin but not bisperoxovanadium 1,10-phenanthroline (bpV(phen)) inhibition of insulin-like growth factor binding protein gene expression. Evidence for MEK-independent activation of mitogen-activated protein kinase by bpV(phen).
The Journal of biological chemistry (1997-01-03)
C J Band, B I Posner
ABSTRACT
The hormonal regulation of insulin-like growth factor binding protein (IGFBP)-1 and -4 mRNA was compared in serum-free primary rat hepatocyte cultures. The combination of dexamethasone and glucagon (Dex/Gluc) strongly increased IGFBP-1 and IGFBP-4 mRNA levels. Insulin suppressed Dex/Gluc-stimulated IGFBP-1 but not IGFBP-4 mRNA levels. In contrast, the peroxovanadium compound, bisperoxovanadium 1,10-phenanthroline (bpV(phen)), completely abrogated Dex/Gluc induction of both IGFBP mRNA species. Wortmannin and rapamycin blocked the inhibitory effect of insulin but not that of bpV(phen) on Dex/Gluc-stimulated IGFBP mRNA. Thus, although phosphatidylinositol 3'-kinase and p70s6k are necessary for insulin-mediated transcriptional inhibition of the IGFBP-1 gene, a signaling pathway, independent of phosphatidyloinositol 3'-kinase and p70s6k, is activated by bpV(phen) and mediates IGFBP-1 as well as IGFBP-4 mRNA inhibition. Mitogen-activated protein (MAP) kinase activity induced by insulin was suppressed to below basal levels in the presence of Dex/Gluc, whereas in response to bpV(phen), MAP kinase activity was high and unaffected by Dex/Gluc, consistent with a role of MAP kinases in bpV(phen)-mediated inhibition of IGFBP mRNA. The specific MAP kinase kinase (MEK) inhibitor, PD98059, inhibited insulin but not bpV(phen)-stimulated MAP kinase activity, suggesting that MAP kinases can be activated in a MEK-independent fashion. Peroxovanadium compounds are strong inhibitors of tyrosine phosphatases, which may inhibit specific tyrosine/threonine phosphatases involved in the negative regulation of MAP kinases.
Sigma-Aldrich
S6 Kinase/Rsk2 Substrate Peptide 2, S6 Kinase/Rsk2 Substrate Peptide 2 primarily used in Kinase Assays.