Merck
Orthogonal reactivity in boryl-substituted organotrifluoroborates.
Journal of the American Chemical Society (2008-11-05)
Gary A Molander, Deidre L Sandrock
ABSTRACT
A method was developed for the hydroboration of alkenyl-containing organotrifluoroborates to generate dibora intermediates. The reactivity differences between organotrifluoroborates and trialkylboranes facilitated the cross-coupling of the borane moiety of these intermediates in a highly chemoselective fashion with aryl halides, leaving the trifluoroborate intact for subsequent transformation. A one-pot hydroboration/two-directional cross-coupling sequence was also demonstrated, providing the fully elaborated products in good yields. These conditions were also amenable in the cross-coupling of trialkylboranes to halo-containing organotrifluoroborates. The stability of the trifluoroborate moiety to these conditions allows simple and efficient strategies for complex molecule construction.
Sigma-Aldrich
Potassium pentafluroroethyltrifluoroborate, 95%