Merck
  • Nickel-catalyzed asymmetric α-arylation and heteroarylation of ketones with chloroarenes: effect of halide on selectivity, oxidation state, and room-temperature reactions.
Nickel-catalyzed asymmetric α-arylation and heteroarylation of ketones with chloroarenes: effect of halide on selectivity, oxidation state, and room-temperature reactions.
Journal of the American Chemical Society (2011-09-16)
Shaozhong Ge, John F Hartwig
ABSTRACT
We report the α-arylation of ketones with a range of aryl chlorides with enantioselectivities from 90 to 99% ee catalyzed by the combination of Ni(COD)(2) and (R)-BINAP and the coupling of ketones with a range of heteroaryl chlorides with enantioselectivities up to 99% ee catalyzed by Ni(COD)(2) and (R)-DIFLUORPHOS. The analogous reactions of bromoarenes occur with much lower enantioselectivities. Mechanistic studies showed that the difference in the rates of decomposition of the arylnickel(II) halide intermediates to {[(R)-BINAP]NiX}(2) likely accounts for the difference in the enantioselectivities of the reactions of bromoarenes and chloroarenes. This catalyst decomposition can be overcome by conducting the reactions with [(R)-BINAP]Ni(η(2)-NC-Ph) (4), which undergoes oxidative addition to haloarenes at room temperature.
Sigma-Aldrich
2-Methyl-1-indanone, 99%