Merck
  • Impact of foliar fungicides on target and non-target soil microbial communities in cucumber crops.
Impact of foliar fungicides on target and non-target soil microbial communities in cucumber crops.
Ecotoxicology and environmental safety (2018-09-25)
Ana Belén López Santísima-Trinidad, María Del Mar Montiel-Rozas, Miguel Ángel Diéz-Rojo, Jose Antonio Pascual, Margarita Ros
ABSTRACT
The application of foliar fungicides to horticultural crops has raised public concerns worldwide. In fact, it has been demonstrated that such fungicides have an impact on non-target microorganisms in the rhizosphere. Fluopyram, triadimenol and penthiopyrad are three broad-spectrum fungicides recommended to control foliar diseases. In our experiment, these fungicides were applied to a cucumber crop to mainly control downy mildew caused by Pseudoperonospora cubensis and grey mold caused by Botrytis cinerea. At the same time, we found that these treatments also controlled other fungal pathogens affecting cucumber crops, particularly penthiopyrad, which was more effective. Once the fungicide application period was over, the effect decreased, although fungicide traces remained in the soil. Furthermore, microbial soil community analysis indicated that both fungicide treatments affect fungal communities to a greater extent than bacterial communities.
Product Description
Supelco
Penthiopyrad, PESTANAL®, analytical standard
Supelco
Fluopyram, PESTANAL®, analytical standard