Merck
  • Entry, Replication, Immune Evasion, and Neurotoxicity of Synthetically Engineered Bat-Borne Mumps Virus.
Entry, Replication, Immune Evasion, and Neurotoxicity of Synthetically Engineered Bat-Borne Mumps Virus.
Cell reports (2018-10-12)
Nadine Krüger, Christian Sauder, Sarah Hüttl, Jan Papies, Kathleen Voigt, Georg Herrler, Kornelia Hardes, Torsten Steinmetzer, Claes Örvell, Jan Felix Drexler, Christian Drosten, Steven Rubin, Marcel Alexander Müller, Markus Hoffmann
ABSTRACT
Bats harbor a plethora of viruses with an unknown zoonotic potential. In-depth functional characterization of such viruses is often hampered by a lack of virus isolates. The genome of a virus closely related to human mumps viruses (hMuV) was detected in African fruit bats, batMuV. Efforts to characterize batMuV were based on directed expression of the batMuV glycoproteins or use of recombinant chimeric hMuVs harboring batMuV glycoprotein. Although these studies provided initial insights into the functionality of batMuV glycoproteins, the host range, replication competence, immunomodulatory functions, virulence, and zoonotic potential of batMuV remained elusive. Here, we report the successful rescue of recombinant batMuV. BatMuV infects human cells, is largely resistant to the host interferon response, blocks interferon induction and TNF-α activation, and is neurotoxic in rats. Anti-hMuV antibodies efficiently neutralize batMuV. The striking similarities between hMuV and batMuV point at the putative zoonotic potential of batMuV.
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HA-Tag antibody, Rabbit Monoclonal, recombinant, expressed in HEK 293 cells, clone RM305, purified immunoglobulin
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution