Merck
  • Genetic and experimental evidence for the involvement of the CD6 lymphocyte receptor in psoriasis.
Genetic and experimental evidence for the involvement of the CD6 lymphocyte receptor in psoriasis.
Cellular & molecular immunology (2017-12-12)
Marta Consuegra-Fernández, Marc Julià, Mario Martínez-Florensa, Fernando Aranda, Cristina Català, Noelia Armiger-Borràs, María-Teresa Arias, Francisca Santiago, Antonio Guilabert, Anna Esteve, Carlos Muñoz, Carlos Ferrándiz, José-Manuel Carrascosa, Edurne Pedrosa, Jorge Romaní, Mercè Alsina, José-Manuel Mascaró-Galy, Francisco Lozano
ABSTRACT
Psoriasis is a chronic inflammatory skin disease with a strong genetic background and is triggered by environmental factors. Available evidence supports CD6, a lymphocyte surface receptor mostly expressed by T cells, as a putative target in autoimmunity. Accordingly, a humanized anti-CD6 antibody has been assayed for the treatment of certain autoimmune disorders, including psoriasis. Here, we present novel evidence in mice and humans for a direct involvement of CD6 in psoriasis pathophysiology. First, an attenuated form of imiquimod-induced psoriasis-like skin inflammation was demonstrated in CD6-deficient mice, as deduced from lower epidermal thickness and local reduced production of pro-inflammatory cytokines, namely, interleukin-17A. Thus, isolated CD4+CD62L+ T cells from CD6-deficient mice displayed decreased in vitro T-helper type 17 polarization. Second, a statistically significant association between CD6 single-nucleotide polymorphisms (rs17824933, rs11230563 and rs12360861) and more severe forms of psoriasis was demonstrated in a cohort of 304 patients at three public hospitals from the metropolitan area of Barcelona. Taken together, these results provide new supportive evidence of the contribution of the CD6 lymphocyte receptor in psoriasis at both experimental and clinical levels.
Sigma-Aldrich
2-(Dimethylamino)ethanethiol hydrochloride, 95%