Merck
  • Phosphonioalkylthiosulfate zwitterions--new masked thiol ligands for the formation of cationic functionalised gold nanoparticles.
Phosphonioalkylthiosulfate zwitterions--new masked thiol ligands for the formation of cationic functionalised gold nanoparticles.
Organic & biomolecular chemistry (2006-11-15)
Yon Ju-Nam, Neil Bricklebank, David W Allen, Philip H E Gardiner, Mark E Light, Michael B Hursthouse
ABSTRACT
We report the synthesis and structural characterisation of a new family of stable phosphonioalkylthiosulfate zwitterions, R3P+ (CH2)nS2O3- (R = Ph or Bu, n = 3,4,6, 8 or 10) which behave as cationic masked thiolate ligands with applications in the functionalisation of gold nanoparticles, having potential as new diagnostic biorecognition systems. The ligands were prepared by treatment of omega-bromoalkylphosphonium salts with sodium thiosulfate. The crystal and molecular structures of the zwitterions (R = Ph, n = 3) and (R = Bu, n = 3) were determined. A series of phosphonioalkanethiolate-capped gold nanoparticles dispersed in water was prepared by borohydride reduction of potassium tetrachloroaurate in the presence of the zwitterions in a dichloromethane-water system. UV-visible spectroscopy and scanning transmission electron-microscopy indicated that capped nanoparticles of ca. 5 nm diameter were present.
Sigma-Aldrich
(6-Bromohexyl)triphenylphosphonium bromide, ≥95%
Pricing and availability is not currently available.