Merck
  • Endometrial progesterone receptor isoforms in women with polycystic ovary syndrome.
Endometrial progesterone receptor isoforms in women with polycystic ovary syndrome.
American journal of translational research (2018-09-14)
Min Hu, Juan Li, Yuehui Zhang, Xin Li, Mats Brännström, Linus R Shao, Håkan Billig
ABSTRACT
Polycystic ovary syndrome (PCOS) affects approximately 4%-18% of all reproductive-aged women and is often accompanied by endometrial progesterone (P4) resistance. Endometrial cells from PCOS patients display increased progesterone receptor (PGR) expression; however, in vivo knockout studies and in vitro experiments indicate the two PGR isoforms are not functionally equivalent. We aimed to compare endometrial PGR isoform expression between non-PCOS and PCOS patients during the proliferative phase. A case-control study. The expression of PGR isoforms (PGRA and PGRB), estrogen receptor alpha (ERα), and markers of cell proliferation was determined by qRT-PCR, Western blot, immunohistochemistry, and immunofluorescence assays. Patients were recruited and diagnosed with PCOS according to the Rotterdam criteria provided by the American Society for Reproductive Medicine and the European Society for Human Reproduction and Embryology. Endometrial biopsy samples were collected from non-PCOS patients with regular menstrual cycles or with hyperplasia (n = 11) and from PCOS patients with or without hyperplasia (n = 14). Although the alteration of PGRB mRNA and protein expression was different, we found that PGRA mRNA and protein expression was higher in PCOS patients than non-PCOS patients. PGRA/B and PGRB were localized in both epithelial and stromal cells, with notable changes in the nuclei of epithelial and stromal cells. A similar expression pattern of ERα, vimentin and Ki-67, in association with an increased PGR expression, was observed in PCOS patients. These results demonstrated that elevated both PGR isoform expression depends on the presence of PCOS, and our data suggest that abnormal regulation of PGR isoforms is a pathological outcome of defective endometrium in PCOS patients.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Cytokeratin Peptide 8 antibody produced in mouse, clone M20, ascites fluid