Merck
  • The Ciliary Machinery Is Repurposed for T Cell Immune Synapse Trafficking of LCK.
The Ciliary Machinery Is Repurposed for T Cell Immune Synapse Trafficking of LCK.
Developmental cell (2018-09-18)
Louise A Stephen, Yasmin ElMaghloob, Michael J McIlwraith, Tamas Yelland, Patricia Castro Sanchez, Pedro Roda-Navarro, Shehab Ismail
ABSTRACT
Upon engagement of the T cell receptor with an antigen-presenting cell, LCK initiates TCR signaling by phosphorylating its activation motifs. However, the mechanism of LCK activation specifically at the immune synapse is a major question. We show that phosphorylation of the LCK activating Y394, despite modestly increasing its catalytic rate, dramatically focuses LCK localization to the immune synapse. We describe a trafficking mechanism whereby UNC119A extracts membrane-bound LCK by sequestering the hydrophobic myristoyl group, followed by release at the target membrane under the control of the ciliary ARL3/ARL13B. The UNC119A N terminus acts as a "regulatory arm" by binding the LCK kinase domain, an interaction inhibited by LCK Y394 phosphorylation, thus together with the ARL3/ARL13B machinery ensuring immune synapse focusing of active LCK. We propose that the ciliary machinery has been repurposed by T cells to generate and maintain polarized segregation of signals such as activated LCK at the immune synapse.
Sigma-Aldrich
Anti-LCK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution