Merck
  • Mutations at multiple CDK phosphorylation consensus sites on Cdt2 increase the affinity of CRL4Cdt2 for PCNA and its ubiquitination activity in S phase.
Mutations at multiple CDK phosphorylation consensus sites on Cdt2 increase the affinity of CRL4Cdt2 for PCNA and its ubiquitination activity in S phase.
Genes to cells : devoted to molecular & cellular mechanisms (2018-02-10)
Kohei Nukina, Akiyo Hayashi, Yasushi Shiomi, Kaoru Sugasawa, Motoaki Ohtsubo, Hideo Nishitani
ABSTRACT
CRL4Cdt2 ubiquitin ligase plays an important role maintaining genome integrity during the cell cycle. A recent report suggested that Cdk1 negatively regulates CRL4Cdt2 activity through phosphorylation of its receptor, Cdt2, but the involvement of phosphorylation remains unclear. To address this, we mutated all CDK consensus phosphorylation sites located in the C-terminal half region of Cdt2 (Cdt2-18A) and examined the effect on substrate degradation. We show that both cyclinA/Cdk2 and cyclinB/Cdk1 phosphorylated Cdt2 in vitro and that phosphorylation was reduced by the 18A mutation both in vitro and in vivo. The 18A mutation increased the affinity of Cdt2 to PCNA, and a high amount of Cdt2-18A was colocalized with PCNA foci during S phase in comparison with Cdt2-WT. Poly-ubiquitination activity to Cdt1 was concomitantly enhanced in cells expressing Cdt2-18A. Other CRL4Cdt2 substrates, Set8 and thymine DNA glycosylase, begin to accumulate around late S phase to G2 phase, but the accumulation was prevented in Cdt2-18A cells. Furthermore, mitotic degradation of Cdt1 after UV irradiation was induced in these cells. Our results suggest that CDK-mediated phosphorylation of Cdt2 inactivates its ubiquitin ligase activity by reducing its affinity to PCNA, an important strategy for regulating the levels of key proteins in the cell cycle.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-SETD8 (hPR-SET7) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit