Merck
  • miR-221/222 Are Involved in Response to Sunitinib Treatment in Metastatic Renal Cell Carcinoma.
miR-221/222 Are Involved in Response to Sunitinib Treatment in Metastatic Renal Cell Carcinoma.
Molecular therapy : the journal of the American Society of Gene Therapy (2015-07-24)
Heba W Z Khella, Henriett Butz, Qiang Ding, Fabio Rotondo, Kenneth R Evans, Peter Kupchak, Moyez Dharsee, Ashraf Latif, Maria D Pasic, Evi Lianidou, Georg A Bjarnason, George M Yousef
ABSTRACT
Sunitinib is a multitargeting tyrosine kinase inhibitor used for metastatic renal cancer. There are no biomarkers that can predict sunitinib response. Such markers are needed to avoid administration of costly medication with side effects to patients who would not benefit from it. We compared global miRNA expression between patients with a short (≤12 months) versus prolonged (>12 months) progression-free survival (PFS) under sunitinib as first-line therapy for metastatic renal cell carcinoma. We identified a number of differentially expressed miRNAs and developed miRNA statistical models that can accurately distinguish between the two groups. We validated our models in the discovery set and an independent set of 57 patients. Target prediction and pathway analysis showed that these miRNAs are involved in vascular endothelial growth factor (VEGF), TGFβ, and mammalian target of rapamycin (mTOR)-mediated signaling and cell-cell communication. We tested the effect of these miRNAs on cellular proliferation and angiogenesis. We validated the negative correlation between miR-221 and its target, VEGFR2.miR-221 overexpression was associated with a poor PFS while its target, VEGFR2 was associated with longer survival. Gain of function experiments showed that miR-221 and miR-222 decreased angiogenesis and cellular proliferation in human umbilical vein endothelial cells (HUVEC) while increasing cellular proliferation in ACHN cells. miRNAs represent potential predictive markers for sunitinib response.
Sigma-Aldrich
Anti-VEGFR2 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.