Merck
  • Nano-targeted induction of dual ferroptotic mechanisms eradicates high-risk neuroblastoma.
Nano-targeted induction of dual ferroptotic mechanisms eradicates high-risk neuroblastoma.
The Journal of clinical investigation (2018-06-26)
Behrouz Hassannia, Bartosz Wiernicki, Irina Ingold, Feng Qu, Simon Van Herck, Yulia Y Tyurina, Hülya Bayır, Behnaz A Abhari, Jose Pedro Friedmann Angeli, Sze Men Choi, Eline Meul, Karen Heyninck, Ken Declerck, Chandra Sekhar Chirumamilla, Maija Lahtela-Kakkonen, Guy Van Camp, Dmitri V Krysko, Paul G Ekert, Simone Fulda, Bruno G De Geest, Marcus Conrad, Valerian E Kagan, Wim Vanden Berghe, Peter Vandenabeele, Tom Vanden Berghe
ABSTRACT
High-risk neuroblastoma is a devastating malignancy with very limited therapeutic options. Here, we identify withaferin A (WA) as a natural ferroptosis-inducing agent in neuroblastoma, which acts through a novel double-edged mechanism. WA dose-dependently either activates the nuclear factor-like 2 pathway through targeting of Kelch-like ECH-associated protein 1 (noncanonical ferroptosis induction) or inactivates glutathione peroxidase 4 (canonical ferroptosis induction). Noncanonical ferroptosis induction is characterized by an increase in intracellular labile Fe(II) upon excessive activation of heme oxygenase-1, which is sufficient to induce ferroptosis. This double-edged mechanism might explain the superior efficacy of WA as compared with etoposide or cisplatin in killing a heterogeneous panel of high-risk neuroblastoma cells, and in suppressing the growth and relapse rate of neuroblastoma xenografts. Nano-targeting of WA allows systemic application and suppressed tumor growth due to an enhanced accumulation at the tumor site. Collectively, our data propose a novel therapeutic strategy to efficiently kill cancer cells by ferroptosis.
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Sigma-Aldrich
ML162, ≥98% (HPLC)
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
Necrostatin-1, Necrostatin-1, CAS 4311-88-0, is a cell-permeable, potent, and selective blocker of necroptosis (EC₅₀ = 494 nM in FADD-deficient Jurkat cells treated with TNF-α).