Merck
  • Self-assembling Properties of an N-Heterocyclic Carbene-based Metallosurfactant: Pd-Coordination Induced Formation of Reactive Interfaces in Water.
Self-assembling Properties of an N-Heterocyclic Carbene-based Metallosurfactant: Pd-Coordination Induced Formation of Reactive Interfaces in Water.
Journal of oleo science (2018-09-04)
Toshiaki Taira, Takaya Yanagimoto, Kenichi Sakai, Hideki Sakai, Akira Endo, Tomohiro Imura
ABSTRACT
In this study, an N-heterocyclic carbene (NHC)-based metallosurfactant (MS), NHC-PdMS, was synthesized, where Pd(II) was bound to the NHC framework via a robust Pd-carbene bond with NEt3 as a co-ligand. Surface tension measurements revealed that the critical micelle concentration (CMC) of NHC-PdMS (1.8×10-4 M) was one order of magnitude lower than that of its MS precursor (imidazolium bromide). Coordination of the MS precursor and NEt3 to Pd(II) also influenced micelle size; the hydrodynamic diameters of NHC-PdMS and the MS precursor were observed to be 25.8±5.6 nm and 2.5±0.3 nm, respectively. Furthermore, small angle X-ray scattering measurements indicated that NHC-PdMS exhibited liquid crystalline behavior above 26 wt%, with a spacing ratio of 1:2:3 for the first, second, and third Bragg peaks. To understand the role of the reactive interface, NHC-PdMS was also applied to aqueous catalytic reactions. Owing to its low CMC value, a catalytic amount of NHC-PdMS (3 mol%) provided the reactive interface, which facilitated the aqueous Mizoroki-Heck reaction of various aryl iodides and styrene in good yields (72-95%). These results suggest that MS formation results in a drastic change in selfassembling properties, which are important for the development of highly reactive chemical interfaces in water.
Sigma-Aldrich
4-Iodobenzoic acid, 98%