Merck
  • Groucho related gene 5 (GRG5) is involved in embryonic and neural stem cell state decisions.
Groucho related gene 5 (GRG5) is involved in embryonic and neural stem cell state decisions.
Scientific reports (2018-09-15)
Konstantina Chanoumidou, Christiana Hadjimichael, Paraskevi Athanasouli, Henrik Ahlenius, Antonis Klonizakis, Christoforos Nikolaou, Elias Drakos, Antonis Kostouros, Irene Stratidaki, Maria Grigoriou, Androniki Kretsovali
ABSTRACT
Groucho related gene 5 (GRG5) is a multifunctional protein that has been implicated in late embryonic and postnatal mouse development. Here, we describe a previously unknown role of GRG5 in early developmental stages by analyzing its function in stem cell fate decisions. By both loss and gain of function approaches we demonstrate that ablation of GRG5 deregulates the Embryonic Stem Cell (ESC) pluripotent state whereas its overexpression leads to enhanced self-renewal and acquisition of cancer cell-like properties. The malignant characteristics of teratomas generated by ESCs that overexpress GRG5 reveal its pro-oncogenic potential. Furthermore, transcriptomic analysis and cell differentiation approaches underline GRG5 as a multifaceted signaling regulator that represses mesendodermal-related genes. When ESCs exit pluripotency, GRG5 promotes neuroectodermal specification via Wnt and BMP signaling suppression. Moreover, GRG5 promotes the neuronal reprogramming of fibroblasts and maintains the self-renewal of Neural Stem Cells (NSCs) by sustaining the activity of Notch/Hes and Stat3 signaling pathways. In summary, our results demonstrate that GRG5 has pleiotropic roles in stem cell biology functioning as a stemness factor and a neural fate specifier.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human AES