Merck
Pd-catalyzed C-N bond formation with heteroaromatic tosylates.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-04-09)
Mette L H Mantel, Anders T Lindhardt, Daniel Lupp, Troels Skrydstrup
ABSTRACT
A protocol for the palladium(0)-catalyzed amidation of heteroaromatic tosylates was successfully developed. The methodology proved to be effective for a variety of heteroaryl tosylates including the pyridine, pyrimidine, quinoline and quinoxaline ring systems. Successful carbon-nitrogen bond formation with these heteroaryl tosylates could be performed with a wide range of primary amides, oxazolidinones, lactams, anilines and indoles, including one cyclic urea. Moreover, this C-N bond forming reaction provided products with high structural diversity. The coupling reaction was also amenable to scale up applications.
Sigma-Aldrich
2-Imidazolidone, 96%