Merck
WNK1 phosphorylates synaptotagmin 2 and modulates its membrane binding.
Molecular cell (2004-09-08)
Byung-Hoon Lee, Xiaoshan Min, Charles J Heise, Bing-E Xu, She Chen, Hongjun Shu, Kate Luby-Phelps, Elizabeth J Goldsmith, Melanie H Cobb
ABSTRACT
WNK (with no lysine [K]) protein kinases were named for their unique active site organization. Mutations in WNK1 and WNK4 cause a familial form of hypertension by undefined mechanisms. Here, we report that WNK1 selectively binds to and phosphorylates synaptotagmin 2 (Syt2) within its calcium binding C2 domains. Endogenous WNK1 and Syt2 coimmunoprecipitate and colocalize on a subset of secretory granules in INS-1 cells. Phosphorylation by WNK1 increases the amount of Ca2+ required for Syt2 binding to phospholipid vesicles; mutation of threonine 202, a WNK1 phosphorylation site, partially prevents this change. These findings suggest that phosphorylation of Syts by WNK1 can regulate Ca2+ sensing and the subsequent Ca2+-dependent interactions mediated by Syt C2 domains. These findings provide a biochemical mechanism that could lead to the retention or insertion of proteins in the plasma membrane. Interruption of this regulatory pathway may disturb membrane events that regulate ion balance.
Sigma-Aldrich
WNK1 (181-507), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.