Merck
WAF1/CIP1 is induced in p53-mediated G1 arrest and apoptosis.
Cancer research (1994-03-01)
W S el-Deiry, J W Harper, P M O'Connor, V E Velculescu, C E Canman, J Jackman, J A Pietenpol, M Burrell, D E Hill, Y Wang
ABSTRACT
The tumor growth suppressor WAF1/CIP1 was recently shown to be induced by p53 and to be a potent inhibitor of cyclin-dependent kinases. In the present studies, we sought to determine the relationship between the expression of WAF1/CIP1 and endogenous regulation of p53 function. WAF1/CIP1 protein was first localized to the nucleus of cells containing wild-type p53 and undergoing G1 arrest. WAF1/CIP1 was induced in wild-type p53-containing cells by exposure to DNA damaging agents, but not in mutant p53-containing cells. The induction of WAF1/CIP1 protein occurred in cells undergoing either p53-associated G1 arrest or apoptosis but not in cells induced to arrest in G1 or to undergo apoptosis through p53-independent mechanisms. DNA damage led to increased levels of WAF1/CIP1 in cyclin E-containing complexes and to an associated decrease in cyclin-dependent kinase activity. These results support the idea that WAF1/CIP1 is a critical downstream effector in the p53-specific pathway of growth control in mammalian cells.
Sigma-Aldrich
p21CIP1, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.